Assay Details
Target Gene Details
Entrez Gene ID: | 57296 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, non-ATPase, 8 |
Gene Aliases: |
6720456J22Rik, AA407360, AL033291, AL033322, AL033323, C76433 |
Location: |
Chr.7:29174187-29180673 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmd8 | NM_026545.3 | 7 | 1054 | NP_080821.3 |
| AK002921.1 | 7 | 893 | BAB22458.2 | |
| AK003436.1 | 7 | 895 | BAB22789.2 | |
| AK020132.1 | 7 | 913 | BAB32006.2 | |
| AK027973.1 | 7 | 858 | BAC25683.1 | |
| AK051257.1 | 7 | 906 | BAC34576.1 | |
| AK150493.1 | 7 | 897 | BAE29609.1 | |
| AK150982.1 | 7 | 861 | BAE30009.1 | |
| AK153329.1 | 7 | 895 | BAE31908.1 | |
| AK159216.1 | 7 | 863 | BAE34905.1 | |
| AK159795.1 | 7 | 879 | BAE35376.1 | |
| AK159926.1 | 7 | 889 | BAE35488.1 | |
| AK166718.1 | 7 | 888 | BAE38968.1 | |
| AK167948.1 | 7 | 863 | BAE39951.1 | |
| AK168886.1 | 6 | 1525 | BAE40703.1 | |
| BC004075.1 | 7 | 872 | AAH04075.1 | |
| BC005717.1 | 7 | 782 | AAH05717.1 | |
| CD774979.1 | 1 | 476 |
More Information
Set Membership: |
|

Genomic Map