Assay Details
Target Gene Details
Entrez Gene ID: | 232784 |
Gene Name: | Zinc finger protein 212 |
Gene Aliases: |
Znf212, mKIAA3011 |
Location: |
Chr.6:47920568-47932639 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp212 | NM_001145881.1 | 5 | 1418 | NP_001139353.1 |
| NM_145576.2 | 5 | 1421 | NP_663551.2 | |
| AK129476.1 | 5 | 1275 | BAC98286.1 | |
| AK218512.1 | ||||
| BC006693.1 | 5 | 1306 | AAH06693.1 | |
| BC064742.1 | 4 | 1246 | AAH64742.1 | |
| BC088737.1 | 5 | 1303 |
More Information
Set Membership: |
|

Genomic Map