Assay Details
Target Gene Details
Entrez Gene ID: | 17984 |
Gene Name: | necdin |
Gene Aliases: |
AI528698, Peg6 |
Location: |
Chr.7:62348277-62349928 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndn | NM_010882.3 | 1 | 831 | NP_035012.2 |
| AK005078.1 | 1 | 789 | BAB23802.1 | |
| AK075729.1 | 1 | 789 | BAC35914.1 | |
| AK075735.1 | 1 | 778 | BAC35917.1 | |
| AK076025.1 | 1 | 787 | BAC36130.1 | |
| AK078129.1 | 1 | 787 | BAC37140.1 | |
| AK135444.1 | 1 | 787 | BAE22535.1 | |
| AK149086.1 | 1 | 787 | BAE28734.1 | |
| BC147249.1 | 1 | 812 | ||
| BC147250.1 | 1 | 812 | ||
| M80840.1 | 1 | 767 | AAA39805.1 |
More Information
Set Membership: |
|

Genomic Map