Assay Details
Target Gene Details
Entrez Gene ID: | 233011 |
Gene Name: | inositol 1,4,5-trisphosphate 3-kinase C |
Gene Aliases: |
9130023N17Rik |
Location: |
Chr.7:27207170-27228597 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpkc | NM_181593.2 | 7 | 2164 | NP_853624.1 |
| AK033605.1 | 7 | 1833 | ||
| AK086754.1 | 1 | 1132 | ||
| AK154888.1 | 7 | 2196 | BAE32904.1 | |
| BC053450.1 | 7 | 2164 | AAH53450.1 |
More Information
Set Membership: |
|

Genomic Map