Assay Details
Target Gene Details
Entrez Gene ID: | 319622 |
Gene Name: | inositol 1,4,5-triphosphate receptor interacting protein-like 2 |
Gene Aliases: |
C130081G24, E030018N11Rik |
Location: |
Chr.7:118485111-118491975 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpripl2 | NM_001033380.3 | 1 | 1149 | NP_001028552.1 |
| AK134593.1 | 1 | 1097 | ||
| AK137678.1 | 1 | 1151 | BAE23457.1 | |
| AK149729.1 | 1 | 1062 | BAE29051.1 | |
| BC150856.1 | 1 | 800 | ||
| BC150864.1 | 1 | 800 |

Genomic Map