Assay Details
Target Gene Details
Entrez Gene ID: | 73919 |
Gene Name: | LYR motif containing 1 |
Gene Aliases: |
1110065L10Rik, 2310004B22Rik, 4930404J24Rik |
Location: |
Chr.7:119895807-119956203 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lyrm1 | NM_001285959.1 | NP_001272888.1 | ||
| NM_001285960.1 | NP_001272889.1 | |||
| NM_029610.3 | 1 | 89 | NP_083886.1 | |
| AK009135.1 | 1 | 92 | BAB26096.1 | |
| AK015083.1 | BAB29706.1 | |||
| BY752674.1 |

Genomic Map