Assay Details
Target Gene Details
Entrez Gene ID: | 114332 |
Gene Name: | lymphatic vessel endothelial hyaluronan receptor 1 |
Gene Aliases: |
1200012G08Rik, Crsbp-1, Lyve-1, Xlkd1 |
Location: |
Chr.7:110850607-110862953 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lyve1 | NM_053247.4 | 6 | 1438 | NP_444477.2 |
| AK004726.2 | 6 | 1438 | BAC25094.1 | |
| AK160857.1 | 5 | 1176 | BAE36050.1 | |
| BC038653.1 | 6 | 1544 | AAH38653.1 | |
| BC038892.1 | 6 | 1544 | AAH38892.1 |
More Information
Set Membership: |
|

Genomic Map