Assay Details
Target Gene Details
Entrez Gene ID: | 72310 |
Gene Name: | natural killer cell group 7 sequence |
Gene Aliases: |
2500004F03Rik |
Location: |
Chr.7:43437138-43438246 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkg7 | NM_024253.4 | 1 | 269 | NP_077215.2 |
| AK010872.1 | 1 | 269 | BAB27236.1 | |
| AK088383.1 | 1 | 278 | BAC40318.1 | |
| BC055882.1 | 1 | 334 | AAH55882.1 |
More Information
Set Membership: |
|

Genomic Map