Assay Details
Target Gene Details
Entrez Gene ID: | 20231 |
Gene Name: | NK1 transcription factor related, locus 2 (Drosophila) |
Gene Aliases: |
Nkx-1.1, Nkx-1.2, Sax1 |
Location: |
Chr.7:132596239-132599637 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx1-2 | NM_009123.2 | 1 | 277 | NP_033149.1 |
| AF222444.1 | 1 | 277 | AAF43671.1 | |
| AF222445.1 | 1 | 278 | AAF43672.1 | |
| AF223361.1 | 1 | 279 | AAF43673.1 | |
| AF223362.1 | AAF43674.1 | |||
| AF223363.1 | AAF43675.1 | |||
| AF223674.1 | AAF43676.1 | |||
| AF223675.1 | AAF43677.1 | |||
| BC139757.1 | 1 | 206 | ||
| X75384.1 | 1 | 382 | CAA53153.1 |
More Information
Set Membership: |
|

Genomic Map