Assay Details
Target Gene Details
Entrez Gene ID: | 22113 |
Gene Name: | pleckstrin homology-like domain, family A, member 2 |
Gene Aliases: |
Ipl, Tssc3 |
Location: |
Chr.7:143501545-143503145 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phlda2 | NM_009434.2 | 2 | 606 | NP_033460.1 |
| AK131929.1 | 2 | 610 | BAE20878.1 | |
| BC019141.1 | 2 | 596 | AAH19141.1 | |
| CF615806.1 | 1 | 161 | ||
| CF617643.1 | 2 | 576 | ||
| Y15443.1 | 2 | 592 | CAA75634.1 |

Genomic Map