Assay Details
Target Gene Details
Entrez Gene ID: | 232337 |
Gene Name: | zinc finger protein 637 |
Gene Aliases: |
AI646709, BC021601, Znf32 |
Location: |
Chr.6:117841156-117845956 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp637 | NM_177684.2 | 3 | 941 | NP_808352.1 |
| AB221671.1 | ||||
| AK146789.1 | 3 | 1074 | BAE27433.1 | |
| BC021601.1 | 3 | 970 | ||
| BC048843.1 | 3 | 941 | AAH48843.1 |
More Information
Set Membership: |
|

Genomic Map