Assay Details
Target Gene Details
Entrez Gene ID: | 243853 |
Gene Name: | fukutin related protein |
Gene Aliases: |
A830029B19Rik, AI842067, AI847300, LGMD1I, MDC1C |
Location: |
Chr.7:16809267-16820668 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkrp | NM_173430.2 | NP_775606.1 | ||
| AJ511806.1 | CAD54301.1 | |||
| AK135089.1 | ||||
| AK156156.1 | ||||
| AK166068.1 | BAE38552.1 | |||
| AK168308.1 | BAE40248.1 | |||
| BC029216.1 | ||||
| BC053072.1 | AAH53072.1 | |||
| BY014040.1 |
Target Gene Details
Entrez Gene ID: | 97387 |
Gene Name: | striatin, calmodulin binding protein 4 |
Gene Aliases: |
C80611, ZIN, zinedin |
Location: |
Chr.7:16815889-16840931 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strn4 | NM_001039878.2 | 1 | 123 | NP_001034967.1 |
| NM_133789.3 | 1 | 123 | NP_598550.2 | |
| AF414080.1 | 1 | 123 | AAL07439.1 |
More Information
Set Membership: |
|

Genomic Map