Assay Details
Target Gene Details
Entrez Gene ID: | 20887 |
Gene Name: | sulfotransferase family 1A, phenol-preferring, member 1 |
Gene Aliases: |
AI266890, PST, ST1A1, Stp, Stp1 |
Location: |
Chr.7:126672865-126676483 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sult1a1 | NM_133670.1 | 7 | 765 | NP_598431.1 |
| AK002700.1 | 7 | 788 | ||
| BC005413.1 | 7 | 765 | AAH05413.1 | |
| L02331.1 | 7 | 781 |
More Information
Set Membership: |
|

Genomic Map