Assay Details
Target Gene Details
Entrez Gene ID: | 232910 |
Gene Name: | adaptor-related protein complex 2, sigma 1 subunit |
Gene Aliases: |
AI043088 |
Location: |
Chr.7:16738444-16749290 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap2s1 | NM_198613.2 | 5 | 608 | NP_941015.2 |
| AK146584.1 | 5 | 609 | BAE27279.1 | |
| AK208745.1 | ||||
| AK212130.1 | ||||
| AK219159.1 | ||||
| BC052499.1 | 6 | 569 | AAH52499.1 |
More Information
Set Membership: |
|

Genomic Map