Assay Details
Target Gene Details
Entrez Gene ID: | 12315 |
Gene Name: | calmodulin 3 |
Gene Aliases: |
CaMA, R75142 |
Location: |
Chr.7:16915379-16924032 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Calm3 | NM_007590.3 | 6 | 1684 | NP_031616.1 |
| AF233282.1 | 1 | 883 | ||
| AK049104.1 | 1 | 420 | ||
| AK083996.1 | 3 | 1290 | BAC39089.2 | |
| AK151610.1 | 6 | 1648 | BAE30549.1 | |
| AK165568.1 | 6 | 1622 | ||
| AK169630.1 | 6 | 1598 | ||
| AK194087.1 | ||||
| BC050926.1 | 6 | 1606 | AAH50926.1 |
More Information
Set Membership: |
|

Genomic Map