Assay Details
Target Gene Details
Entrez Gene ID: | 16985 |
Gene Name: | lymphocyte specific 1 |
Gene Aliases: |
Lsp-1, WP34, p50, pp52 |
Location: |
Chr.7:142460812-142494868 on Build GRCm38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lsp1 | NM_001136071.2 | NP_001129543.1 | ||
| NM_001271508.1 | NP_001258437.1 | |||
| NM_001271509.1 | NP_001258438.1 | |||
| NM_001271510.1 | NP_001258439.1 | |||
| NM_001271523.1 | NP_001258452.1 | |||
| NM_019391.3 | NP_062264.1 | |||
| AK003081.1 | ||||
| AK075783.1 | ||||
| BC003796.1 | AAH03796.1 | |||
| D49691.1 | BAA08541.1 | |||
| M89956.1 | AAB48537.1 | |||
| M90316.1 | AAA65108.1 |
Target Gene Details
Entrez Gene ID: | 677289 |
Gene Name: | proline rich 33 |
Gene Aliases: |
ENSMUSG00000043795, Gm14492, OTTMUSG00000016823 |
Location: |
Chr.7:142491080-142506771 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prr33 | NR_033261.1 | 3 | 990 | |
| AK082720.1 | 3 | 991 | BAC38585.1 |
More Information
Set Membership: |
|

Genomic Map