Assay Details
Target Gene Details
Entrez Gene ID: | 12862 |
Gene Name: | cytochrome c oxidase subunit VIa polypeptide 2 |
Gene Aliases: |
CoxVIaH, VIaH |
Location: |
Chr.7:128205435-128206366 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox6a2 | NM_009943.2 | 3 | 343 | NP_034073.2 |
| AK002506.1 | 3 | 294 | BAB22151.1 | |
| BC028514.1 | 3 | 314 | AAH28514.1 | |
| BF472154.1 | 3 | 226 | ||
| BU842968.1 | 3 | 343 | ||
| U08439.1 | 3 | 329 | AAA17835.1 |
More Information
Set Membership: |
|

Genomic Map