Assay Details
Target Gene Details
Entrez Gene ID: | 69546 |
Gene Name: | mitogen-activated protein kinase 1 interacting protein 1 |
Gene Aliases: |
2310009E07Rik, AU043776, MISS |
Location: |
Chr.7:138835818-138846267 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk1ip1 | NM_001045483.1 | 1 | 109 | NP_001038948.1 |
| NM_027115.2 | 1 | 109 | NP_081391.1 | |
| AK009250.1 | 1 | 100 | BAB26168.1 | |
| BE623651.1 | 1 | 85 | ||
| BY706993.1 | 1 | 109 |

Genomic Map