Assay Details
Target Gene Details
Entrez Gene ID: | 78748 |
Gene Name: | Ras association (RalGDS/AF-6) domain family (N-terminal) member 10 |
Gene Aliases: |
4632411J06Rik |
Location: |
Chr.7:112953962-112957458 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf10 | NM_175279.3 | 1 | 583 | NP_780488.2 |
| AK019489.1 | ||||
| AK031660.1 | 1 | 584 | BAC27498.1 | |
| AK046433.1 | 1 | 578 | BAC32726.1 | |
| BB621318.1 | 1 | 584 | ||
| BC028854.1 | 1 | 159 | AAH28854.1 | |
| BC125291.1 | 1 | 399 | ||
| BC125293.1 | 1 | 399 |
More Information
Set Membership: |
|

Genomic Map