Assay Details
Target Gene Details
Entrez Gene ID: | 55942 |
Gene Name: | SERTA domain containing 1 |
Gene Aliases: |
1110032C13Rik, AV000455, AW456625, Sei-1, Sei1, TRIP-Br1 |
Location: |
Chr.7:27486953-27490316 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sertad1 | NM_018820.4 | 2 | 839 | NP_061290.1 |
| AF218291.1 | 2 | 746 | AAF27653.1 | |
| AF366401.1 | 2 | 882 | AAK52830.1 | |
| AK002324.1 | 2 | 840 | BAB22013.1 | |
| AK004022.1 | 2 | 839 | BAB23130.1 | |
| AK008303.1 | 2 | 840 | BAB25588.1 | |
| BC016077.1 | 2 | 799 | AAH16077.1 |
More Information
Set Membership: |
|

Genomic Map