Assay Details
Target Gene Details
Entrez Gene ID: | 76572 |
Gene Name: | RNA binding motif protein, X-linked-like 2 |
Gene Aliases: |
1700012H05Rik, Hnrnpg-t |
Location: |
Chr.7:107209445-107210916 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbmxl2 | NM_029660.2 | 1 | 1224 | NP_083936.1 |
| AK005907.1 | 1 | 1225 | BAB24311.1 |

Genomic Map