Assay Details
Target Gene Details
Entrez Gene ID: | 244218 |
Gene Name: | cardiotrophin 2 |
Gene Aliases: |
Gm494, NP |
Location: |
Chr.7:127717816-127725616 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctf2 | NM_198858.1 | 3 | 780 | NP_942155.1 |
| AB125661.1 | 3 | 780 | BAD01485.1 | |
| BC120763.1 | ||||
| BC120765.1 |

Genomic Map