Assay Details
Target Gene Details
Entrez Gene ID: | 13033 |
Gene Name: | cathepsin D |
Gene Aliases: |
CD, CatD |
Location: |
Chr.7:142375916-142387870 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 13033 |
Gene Name: | cathepsin D |
Gene Aliases: |
CD, CatD |
Location: |
Chr.7:142375916-142387870 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |