Assay Details
Target Gene Details
Entrez Gene ID: | 14082 |
Gene Name: | Fas (TNFRSF6)-associated via death domain |
Gene Aliases: |
Mort1/FADD |
Location: |
Chr.7:144578323-144582436 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fadd | NM_010175.5 | 2 | 2025 | NP_034305.1 |
| AK084808.1 | 2 | 2026 | BAC39283.1 | |
| AK169798.1 | 2 | 2053 | BAE41374.1 |

Genomic Map