Assay Details
Target Gene Details
Entrez Gene ID: | 101612 |
Gene Name: | glutamate-rich WD repeat containing 1 |
Gene Aliases: |
A301, AI504353, Grwd, Wdr28 |
Location: |
Chr.7:45825223-45830796 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grwd1 | NM_153419.2 | 7 | 1464 | NP_700468.2 |
| AB001539.1 | 8 | 1536 | BAC66461.1 | |
| AK077611.1 | 7 | 1470 | BAC36899.1 | |
| AK210248.1 | ||||
| BC008121.1 | 7 | 1441 | AAH08121.1 | |
| BC028896.1 | 6 | 1333 | AAH28896.1 | |
| BC083143.1 | 7 | 1452 | AAH83143.1 | |
| DT904438.1 | 1 | 279 |

Genomic Map