Assay Details
Target Gene Details
Entrez Gene ID: | 192658 |
Gene Name: | ret finger protein-like 4 |
Gene Aliases: |
D7Ertd486e, Rfpl4a |
Location: |
Chr.7:5109787-5116911 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfpl4 | NM_001145013.1 | 3 | 711 | NP_001138485.1 |
| NM_138954.3 | 3 | 943 | NP_620404.1 | |
| AK136022.1 | 3 | 712 | BAE22777.1 | |
| AK139569.1 | 3 | 945 | ||
| AY070253.1 | 3 | 933 | AAL55432.1 | |
| BC069834.1 | 3 | 697 | AAH69834.1 |

Genomic Map