Assay Details
Target Gene Details
Entrez Gene ID: | 12476 |
Gene Name: | CD151 antigen |
Gene Aliases: |
PETA-3, SFA-1, Tspan24 |
Location: |
Chr.7:141467362-141471481 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd151 | NM_001111049.1 | 8 | 1159 | NP_001104519.1 |
| NM_001111050.1 | 7 | 1034 | NP_001104520.1 | |
| NM_009842.3 | 8 | 1277 | NP_033972.2 | |
| AK178760.1 | 1 | 165 | ||
| AK179098.1 | 1 | 94 | ||
| AK183639.1 | 1 | 94 | ||
| AK187557.1 | 1 | 160 | ||
| AK206440.1 | 1 | 94 | ||
| AK207853.1 | 1 | 53 | ||
| AK209805.1 | 1 | 94 | ||
| AK212781.1 | 1 | 104 | ||
| AK214308.1 | 1 | 94 | ||
| AK214748.1 | 1 | 114 | ||
| AK218332.1 | 1 | 67 | ||
| BC012236.1 | 7 | 956 | AAH12236.1 | |
| D89290.1 | 9 | 1204 | BAA22447.1 | |
| U89772.1 | 7 | 1001 | AAC25976.1 |

Genomic Map