Assay Details
Target Gene Details
Entrez Gene ID: | 13089 |
Gene Name: | cytochrome P450, family 2, subfamily b, polypeptide 13 |
Gene Aliases: |
- |
Location: |
Chr.7:26061344-26096197 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2b13 | NM_007813.2 | 9 | 1783 | NP_031839.2 |
More Information
Set Membership: |
|

Genomic Map