Assay Details
Target Gene Details
Entrez Gene ID: | 13107 |
Gene Name: | cytochrome P450, family 2, subfamily f, polypeptide 2 |
Gene Aliases: |
Cyp2f |
Location: |
Chr.7:27119955-27133660 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2f2 | NM_007817.2 | 10 | 1668 | NP_031843.2 |
| BC011089.1 | 10 | 1639 | AAH11089.1 | |
| BC024742.1 | 10 | 1649 | AAH24742.1 | |
| CK626215.1 | 1 | 175 | ||
| M77497.1 | 10 | 1648 | AAA37517.1 |
More Information
Set Membership: |
|

Genomic Map