Assay Details
Target Gene Details
Entrez Gene ID: | 73813 |
Gene Name: | family with sequence similarity 83, member E |
Gene Aliases: |
4930403C10Rik |
Location: |
Chr.7:45721220-45729492 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam83e | NM_001033170.4 | 4 | 2026 | NP_001028342.2 |
| AK015058.1 | 2 | 352 | ||
| BC042793.1 | 4 | 873 | AAH42793.1 |
Target Gene Details
Entrez Gene ID: | 54200 |
Gene Name: | sulfotransferase family, cytosolic, 2B, member 1 |
Gene Aliases: |
AI326997, BB173635, ST2B1, SULT2B |
Location: |
Chr.7:45721174-45759555 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sult2b1 |

Genomic Map