Assay Details
Target Gene Details
Entrez Gene ID: | 233204 |
Gene Name: | TBC1 domain family, member 17 |
Gene Aliases: |
BC017607 |
Location: |
Chr.7:44840776-44849079 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d17 | NM_001042655.1 | 17 | 2390 | NP_001036120.1 |
| AK046883.1 | 16 | 2327 | BAC32907.1 | |
| AK083153.1 | 17 | 2208 |
More Information
Set Membership: |
|

Genomic Map