Assay Details
Target Gene Details
Entrez Gene ID: | 107503 |
Gene Name: | activating transcription factor 5 |
Gene Aliases: |
AFTA, Atf7, Atfx, ODA-10 |
Location: |
Chr.7:44812256-44816658 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf5 | NM_030693.2 | 2 | 232 | NP_109618.1 |
| NR_033136.1 | 2 | 312 | ||
| AF310624.1 | 2 | 253 | AAL55820.1 | |
| AF312938.1 | 2 | 208 | AAG34160.1 | |
| AF375475.1 | 2 | 237 | AAK54640.1 | |
| AF375476.1 | 2 | 232 | AAK54641.1 | |
| AK131939.1 | 2 | 253 | ||
| AK146479.1 | 2 | 254 | BAE27202.1 | |
| AK159624.1 | 2 | 257 | ||
| BC010195.1 | 1 | 65 | AAH10195.1 | |
| BC092068.1 | 2 | 237 | AAH92068.1 | |
| BY307992.1 | 2 | 313 |
More Information
Set Membership: |
|

Genomic Map