Assay Details
Target Gene Details
Entrez Gene ID: | 74134 |
Gene Name: | cytochrome P450, family 2, subfamily s, polypeptide 1 |
Gene Aliases: |
1200011C15Rik, AU041727, C79779 |
Location: |
Chr.7:25802476-25816530 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2s1 | NM_028775.3 | 9 | 1984 | NP_083051.1 |
| AK004699.2 | 9 | 1984 | BAB23484.1 | |
| AK087069.1 | 9 | 1972 | BAC39794.1 | |
| AK162947.1 | 8 | 2524 | ||
| BC034202.1 | 6 | 1424 | AAH34202.1 | |
| BC064733.1 | 9 | 2001 | AAH64733.1 |
More Information
Set Membership: |
|

Genomic Map