Assay Details
Target Gene Details
Entrez Gene ID: | 19158 |
Gene Name: | cytohesin 2 |
Gene Aliases: |
ARNO, CLM2, Pscd2 |
Location: |
Chr.7:45806637-45814316 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyth2 | NM_001112701.1 | 12 | 1915 | NP_001106171.1 |
| NM_011181.3 | 13 | 1918 | NP_035311.1 | |
| AK045451.1 | 13 | 1887 | BAC32376.1 |
More Information
Set Membership: |
|

Genomic Map