Assay Details
Target Gene Details
Entrez Gene ID: | 21849 |
Gene Name: | tripartite motif-containing 28 |
Gene Aliases: |
AA408787, KAP-1, KRIP-1, MommeD9, Tif1b, Tif1beta |
Location: |
Chr.7:13024152-13031032 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim28 | NM_011588.3 | 1 | 196 | NP_035718.2 |
| AK131807.1 | 1 | 250 | ||
| BC089337.1 | 1 | 196 | AAH89337.1 | |
| U67303.1 | 1 | 188 | AAB17272.1 |

Genomic Map