Assay Details
Target Gene Details
Entrez Gene ID: | 109815 |
Gene Name: | VCP-interacting membrane protein |
Gene Aliases: |
1500011E07Rik, C78786, H-4, H-47, H4, H47, Sels |
Location: |
Chr.7:66079649-66089405 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vimp | NM_024439.3 | 6 | 767 | NP_077759.3 |
| AF335543.1 | 6 | 721 | AAK07659.1 | |
| AK005204.1 | 6 | 750 | BAB23882.1 | |
| AK131659.1 | ||||
| AK214656.1 | 1 | 142 | ||
| BC011091.1 | 6 | 757 | AAH11091.1 |

Genomic Map