Assay Details
Target Gene Details
Entrez Gene ID: | 110959 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 19 |
Gene Aliases: |
D7Rp2, D7Rp2-r, D7Rp2-s, D7Rp2e, RP2-r, RP2-s |
Location: |
Chr.7:35547185-35556417 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt19 | NM_033080.2 | 1 | 262 | NP_149071.2 |
| AK011801.1 | 1 | 117 | ||
| BC138674.1 | 1 | 185 | ||
| BC138675.1 | 1 | 185 | ||
| CJ063443.1 | 1 | 263 |

Genomic Map