Assay Details
Target Gene Details
Entrez Gene ID: | 18226 |
Gene Name: | nucleoporin 62 |
Gene Aliases: |
AA589433, AI426861, AI790512, AU045898, D7Ertd649e, Nupc1, p62 |
Location: |
Chr.7:44816420-44830807 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup62 | NM_053074.1 | 2 | 1215 | NP_444304.1 |
| AK147894.1 | 2 | 1244 | BAE28211.1 | |
| AK149637.1 | 2 | 1266 | BAE28999.1 | |
| AK150087.1 | 2 | 1236 | BAE29296.1 | |
| AK155059.1 | 2 | 1240 | BAE33019.1 | |
| AK166621.1 | 2 | 1383 | BAE38900.1 | |
| AK168166.1 | 2 | 1237 | BAE40127.1 | |
| AK168486.1 | 2 | 1248 | BAE40373.1 | |
| AK169178.1 | 2 | 1259 | BAE40956.1 | |
| AK171455.1 | 2 | 1550 | BAE42464.1 | |
| AK171611.1 | 2 | 1548 | BAE42562.1 | |
| AK188830.1 | ||||
| AK206351.1 | ||||
| BC005784.1 | 2 | 1215 | AAH05784.1 | |
| BC089317.1 | 2 | 1296 | AAH89317.1 | |
| S59342.1 | 2 | 1213 | AAB19953.1 |
Target Gene Details
Entrez Gene ID: | 100328588 |
Gene Name: | Nup62-Il4i1 protein |
Gene Aliases: |
- |
Location: |
Chr.7:44816370-44840804 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup62-il4i1 | NM_001171024.1 | NP_001164495.1 | ||
| AK133082.1 | BAE21502.1 | |||
| BY087056.1 | ||||
| BY088421.1 | ||||
| CJ047381.1 |
More Information
Set Membership: |
|

Genomic Map