Assay Details
Target Gene Details
Entrez Gene ID: | 267019 |
Gene Name: | ribosomal protein S15A |
Gene Aliases: |
A630031B11Rik |
Location: |
Chr.7:118104374-118116147 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps15a | NM_170669.2 | 4 | 1127 | NP_733769.1 |
| AK032522.1 | 5 | 1167 | BAC27909.1 | |
| BC054792.1 | 4 | 1127 | AAH54792.1 |
More Information
Set Membership: |
|

Genomic Map