Assay Details
Target Gene Details
Entrez Gene ID: | 20130 |
Gene Name: | Harvey rat sarcoma oncogene, subgroup R |
Gene Aliases: |
AI573426 |
Location: |
Chr.7:45018007-45021647 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rras | NM_009101.2 | 6 | 697 | NP_033127.1 |
| AK155855.1 | 6 | 727 | BAE33462.1 | |
| BC009105.1 | 6 | 714 | AAH09105.1 | |
| BC110562.1 | 6 | 699 | AAI10563.1 | |
| BC110563.1 | 6 | 699 | AAI10564.1 | |
| BC145932.1 | 6 | 743 | ||
| BC145934.1 | 6 | 743 | ||
| M21019.1 | 6 | 699 | AAA40038.1 |
More Information
Set Membership: |
|

Genomic Map