Assay Details
Target Gene Details
Entrez Gene ID: | 21804 |
Gene Name: | transforming growth factor beta 1 induced transcript 1 |
Gene Aliases: |
ARA55, Hic5, TSC-5, hic-5 |
Location: |
Chr.7:128246799-128255699 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfb1i1 | NM_001289550.1 | 11 | 1709 | NP_001276479.1 |
| NM_001289551.1 | 11 | 1705 | NP_001276480.1 | |
| NM_001289552.1 | 10 | 1566 | NP_001276481.1 | |
| NM_001289553.1 | 10 | 1593 | NP_001276482.1 | |
| AK003170.1 | 11 | 1543 | ||
| AK041233.1 | 11 | 1538 | ||
| AK085042.1 | 10 | 1436 | ||
| AK087114.1 | 11 | 1540 | ||
| BC002049.1 | 11 | 1543 | AAH02049.1 | |
| BC056362.1 | 10 | 2818 | AAH56362.1 | |
| DQ143891.1 | 10 | 1594 | AAZ82195.1 | |
| DQ143892.1 | 9 | 1380 | AAZ82196.1 | |
| DQ143893.1 | 9 | 1387 | AAZ82197.1 | |
| DQ143894.1 | 9 | 1382 | AAZ82198.1 | |
| DQ143895.1 | 9 | 1798 | AAZ82199.1 | |
| DQ143896.1 | 9 | 1759 | AAZ82200.1 | |
| DQ143897.1 | 9 | 1837 | AAZ82201.1 | |
| DQ143898.1 | 9 | 1786 | AAZ82202.1 | |
| DQ143899.1 | 8 | 1913 | AAZ82203.1 | |
| DQ143900.1 | 10 | 1606 | AAZ82204.1 | |
| L22482.1 | 10 | 1670 | AAA62226.1 |
More Information
Set Membership: |
|

Genomic Map