Assay Details
Target Gene Details
Entrez Gene ID: | 107503 |
Gene Name: | activating transcription factor 5 |
Gene Aliases: |
AFTA, Atf7, Atfx, ODA-10 |
Location: |
Chr.7:44812256-44816658 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf5 | NM_030693.2 | 3 | 896 | NP_109618.1 |
| NR_033136.1 | 3 | 976 | ||
| AB012276.1 | BAA25313.1 | |||
| AF310624.1 | 3 | 917 | AAL55820.1 | |
| AF312938.1 | 3 | 872 | AAG34160.1 | |
| AF375475.1 | 3 | 901 | AAK54640.1 | |
| AF375476.1 | 3 | 896 | AAK54641.1 | |
| AK131939.1 | 3 | 917 | ||
| AK146479.1 | 3 | 918 | BAE27202.1 | |
| AK159624.1 | 3 | 920 | ||
| AK160085.1 | 1 | 321 | BAE35616.1 | |
| AY024362.1 | 1 | 405 | AAK00584.1 | |
| BC010195.1 | 2 | 729 | AAH10195.1 | |
| BC092068.1 | 3 | 901 | AAH92068.1 |
More Information
Set Membership: |
|

Genomic Map