Assay Details
Target Gene Details
Entrez Gene ID: | 13170 |
Gene Name: | D site albumin promoter binding protein |
Gene Aliases: |
- |
Location: |
Chr.7:45705095-45710203 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbp | NM_016974.3 | 1 | 174 | NP_058670.2 |
| AK140243.1 | 1 | 51 | BAE24294.1 | |
| BC018323.1 | 1 | 137 | AAH18323.1 | |
| BC084676.1 | 1 | 152 | AAH84676.1 | |
| CK619069.1 | 1 | 152 | ||
| CX217039.1 | 1 | 297 |
More Information
Set Membership: |
|

Genomic Map