Assay Details
Target Gene Details
Entrez Gene ID: | 13170 |
Gene Name: | D site albumin promoter binding protein |
Gene Aliases: |
- |
Location: |
Chr.7:45705095-45710203 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbp | NM_016974.3 | 2 | 726 | NP_058670.2 |
| AK140243.1 | 2 | 603 | BAE24294.1 | |
| BC018323.1 | 2 | 689 | AAH18323.1 | |
| BC084676.1 | AAH84676.1 |
More Information
Set Membership: |
|

Genomic Map