Assay Details
Target Gene Details
Entrez Gene ID: | 13170 |
Gene Name: | D site albumin promoter binding protein |
Gene Aliases: |
- |
Location: |
Chr.7:45705095-45710203 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbp | NM_016974.3 | 4 | 1586 | NP_058670.2 |
| AK140243.1 | 4 | 1463 | BAE24294.1 | |
| AK192990.1 | 1 | 116 |
Target Gene Details
Entrez Gene ID: | 56632 |
Gene Name: | sphingosine kinase 2 |
Gene Aliases: |
C76851 |
Location: |
Chr.7:45709463-45718002 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sphk2 | NM_001172561.1 | 6 | 2833 | NP_001166032.1 |
| NM_020011.5 | 6 | 3142 | NP_064395.2 | |
| NM_203280.3 | 7 | 2904 | NP_975009.1 | |
| AK016616.1 | 3 | 1922 | ||
| AK154321.1 | 6 | 2570 | BAE32512.1 | |
| AK208751.1 | ||||
| AK218410.1 |
More Information
Set Membership: |
|

Genomic Map