Assay Details
Target Gene Details
Entrez Gene ID: | 233079 |
Gene Name: | free fatty acid receptor 2 |
Gene Aliases: |
GPCR43, Gpr43 |
Location: |
Chr.7:30818032-30823797 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ffar2 | NM_146187.4 | 1 | 54 | NP_666299.1 |
| AK078861.1 | 1 | 54 | BAC37425.1 |

Genomic Map