Assay Details
Target Gene Details
Entrez Gene ID: | 233079 |
Gene Name: | free fatty acid receptor 2 |
Gene Aliases: |
GPCR43, Gpr43 |
Location: |
Chr.7:30818032-30823797 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ffar2 | NM_001168509.1 | 3 | 683 | NP_001161981.1 |
| NM_001168510.1 | 3 | 620 | NP_001161982.1 | |
| NM_001168511.1 | 3 | 627 | NP_001161983.1 | |
| NM_001168512.1 | 2 | 773 | NP_001161984.1 | |
| NM_146187.4 | 3 | 1006 | NP_666299.1 | |
| AF545043.1 | 3 | 557 | AAO16236.1 | |
| AK050524.1 | 1 | 638 | BAC34306.1 | |
| AK078861.1 | 3 | 1006 | BAC37425.1 | |
| BC019570.1 | 1 | 683 | AAH19570.1 | |
| CX228700.1 |

Genomic Map