Assay Details
Target Gene Details
Entrez Gene ID: | 15373 |
Gene Name: | H6 homeobox 3 |
Gene Aliases: |
Nkx-5.1, Nkx5-1, Nkx5.1 |
Location: |
Chr.7:131541138-131546027 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmx3 | NM_008257.3 | 1 | 66 | NP_032283.3 |
| BC139482.1 | 1 | 162 | ||
| BC139483.1 | 1 | 162 |
More Information
Set Membership: |
|

Genomic Map