Assay Details
Target Gene Details
Entrez Gene ID: | 70394 |
Gene Name: | kaptin |
Gene Aliases: |
2310042D10Rik, 2E4, C030013F01Rik |
Location: |
Chr.7:16119876-16127516 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kptn | NM_133727.2 | 1 | 259 | NP_598488.2 |
| AK009746.1 | 1 | 241 | ||
| AK081226.1 | 1 | 251 | ||
| BC018316.1 | 1 | 227 | AAH18316.1 | |
| BY269246.1 | 1 | 260 |

Genomic Map