Assay Details
Target Gene Details
Entrez Gene ID: | 233222 |
Gene Name: | MAS-related GPR, member A3 |
Gene Aliases: |
Mrga3 |
Location: |
Chr.7:47588950-47601372 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrgpra3 | NM_153067.2 | 2 | 896 | NP_694707.2 |
| AY042193.1 | 2 | 911 | AAK91789.1 |
More Information
Set Membership: |
|

Genomic Map